Skip to Content
MilliporeSigma
All Photos(2)

Documents

720011

Sigma-Aldrich

Potassium 2-iodo-5-methylbenzenesulfonate

greener alternative

Synonym(s):

Pre-IBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6IKO3S
CAS Number:
Molecular Weight:
336.19
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

288-292 °C

greener alternative category

storage temp.

2-8°C

SMILES string

[K+].Cc1ccc(I)c(c1)S([O-])(=O)=O

InChI

1S/C7H7IO3S.K/c1-5-2-3-6(8)7(4-5)12(9,10)11;/h2-4H,1H3,(H,9,10,11);/q;+1/p-1

InChI key

LFQHEPUFDZVNHD-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

  • Catalyst for alcohol oxidation
Catalyst for greener oxidation of alcohols using Oxone as the terminal oxidant. Alternative to stoichiometric hypervalent iodine oxidants.

2-Iodoxybenzenesulfonic Acid as an Extremely Active Catalyst for the Selective Oxidation of Alcohols to Aldehydes, Ketones, Carboxylic Acids, and Enones with Oxone

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service