Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

719609

Sigma-Aldrich

2-Chloro-6-(tributylstannyl)pyridine

Synonym(s):

2-(Tributylstannyl)-6-chloropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30ClNSn
CAS Number:
Molecular Weight:
402.59
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

density

1.191 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cccc(Cl)n1

InChI

1S/C5H3ClN.3C4H9.Sn/c6-5-3-1-2-4-7-5;3*1-3-4-2;/h1-3H;3*1,3-4H2,2H3;

InChI key

SRBYIFPGENOZFS-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service