Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718335

Sigma-Aldrich

2-Aminoquinoline

97%

Synonym(s):

2-Quinolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

126-131 °C

SMILES string

Nc1ccc2ccccc2n1

InChI

1S/C9H8N2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H,(H2,10,11)

InChI key

GCMNJUJAKQGROZ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaoxue Lv et al.
Molecular carcinogenesis, 58(5), 794-807 (2019-01-08)
Core fucosylation catalyzed by core fucosyltransferase (Fut8) contributes to the progressions of epithelial ovarian cancer (EOC). Copper transporter 1 (CTR1), which contains one N-glycan on Asn15 , mediates cellular transport of cisplatin (cDDP), and plays an important role in the
Chenyang He et al.
The Analyst, 145(13), 4504-4511 (2020-05-16)
Dabrafenib is one of the most widely used of the new generation of targeted anti-cancer drugs. However, its therapeutic window varies for different patients and so there is an unmet need for methods to monitor the dose of drug which
Raquel de Meneses Santos et al.
Bioorganic & medicinal chemistry, 23(15), 4390-4396 (2015-06-28)
In this paper, we describe the synthesis and pharmacological evaluation of a series of 4-aminoquinolines. The compounds were characterised and tested in models of pain and inflammation, using the writhing test with acetic acid, formalin test, peritonitis test by zymosan

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service