Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718092

Sigma-Aldrich

3-Phenylisoxazole-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

178-182 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(no1)-c2ccccc2

InChI

1S/C10H7NO3/c12-10(13)9-6-8(11-14-9)7-4-2-1-3-5-7/h1-6H,(H,12,13)

InChI key

YGTFDJRCCBKLDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service