Skip to Content
MilliporeSigma
All Photos(1)

Documents

717789

Sigma-Aldrich

[((1R,2R)-2-Amino-1,2-diphenylethyl)(p-tosyl)amido](p-cymene)ruthenium(II) tetrafluoroborate

97%

Synonym(s):

RuBF4(p-cymene)[(R,R)-TS-DPEN]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35BF4N2O2RuS
Molecular Weight:
687.56
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

207-212 °C

SMILES string

F[B-](F)(F)F.C[C]1[CH][CH][C]([CH][CH]1)C(C)C.Cc2ccc(cc2)S(=O)(=O)N([Ru+])[C@@H]([C@H](N)c3ccccc3)c4ccccc4

InChI

1S/C21H21N2O2S.C10H14.BF4.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-8(2)10-6-4-9(3)5-7-10;2-1(3,4)5;/h2-15,20-21H,22H2,1H3;4-8H,1-3H3;;/q-1;;-1;+2/t20-,21-;;;/m1.../s1

InChI key

BMEOXNQRLZPIDE-AGEKDOICSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service