Skip to Content
MilliporeSigma
All Photos(1)

Documents

717452

Sigma-Aldrich

Tris(triphenylsiloxy)vanadium oxide

95%

Synonym(s):

NSC 349960, Oxotris(triphenylsilanolato) vanadium, Triphenylsilanol vanadate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H45O4Si3V
CAS Number:
Molecular Weight:
893.13
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

95%

form

powder

reaction suitability

core: vanadium
reagent type: catalyst

mp

224-226 °C

SMILES string

O=[V](O[Si](c1ccccc1)(c2ccccc2)c3ccccc3)(O[Si](c4ccccc4)(c5ccccc5)c6ccccc6)O[Si](c7ccccc7)(c8ccccc8)c9ccccc9

InChI

1S/3C18H15OSi.O.V/c3*19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;;/q3*-1;;+3

InChI key

WSQFWIWHGOUAEN-UHFFFAOYSA-N

Application

Catalyst for:
  • Dual catalyst system for Meyer-Schuster rearrangement
  • Esterification
  • Addition of propargyl alcohols and imines or aldehydes
  • Photochemical polymerization of Me methacylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service