Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

716766

Sigma-Aldrich

(2R,5R)-1-(((2R,5R)-2,5-Diphenylpyrrolidin-1-yl)methylene)-2,5-diphenylpyrrolidinium tetrafluoroborate

Synonym(s):

Bis[(2R,5R)-diphenyl-1-pyrrolidinyl]methylium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H33BF4N2
CAS Number:
Molecular Weight:
544.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]22/D +144.0°, c = 0.5 in dichloromethane

mp

>300 °C

functional group

phenyl

SMILES string

F[B-](F)(F)F.C1C[C@@H](N(\C=[N+]2/[C@H](CC[C@@H]2c3ccccc3)c4ccccc4)[C@H]1c5ccccc5)c6ccccc6

InChI

1S/C33H33N2.BF4/c1-5-13-26(14-6-1)30-21-22-31(27-15-7-2-8-16-27)34(30)25-35-32(28-17-9-3-10-18-28)23-24-33(35)29-19-11-4-12-20-29;2-1(3,4)5/h1-20,25,30-33H,21-24H2;/q+1;-1/t30-,31-,32-,33-;/m1./s1

InChI key

RKLHUHRTFDXZEQ-ZNOPIPGTSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service