Skip to Content
MilliporeSigma
All Photos(1)

Documents

716693

Sigma-Aldrich

(1S,2S)-2-(Diphenylphosphino)cyclohexylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22NP
CAS Number:
Molecular Weight:
283.35
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]22/D +103.0°, c = 0.5 in dichloromethane

mp

51-56 °C

SMILES string

N[C@H]1CCCC[C@@H]1P(c2ccccc2)c3ccccc3

InChI

1S/C18H22NP/c19-17-13-7-8-14-18(17)20(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-6,9-12,17-18H,7-8,13-14,19H2/t17-,18-/m0/s1

InChI key

ZATLZEHZPXYMFE-ROUUACIJSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service