Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

716618

Sigma-Aldrich

4′-Methoxy-2-chloroacetophenone

97%

Synonym(s):

2-Chloro-1-(4-methoxyphenyl)ethanone, 4-(Chloroacetyl)anisole, 4-Methoxyphenacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

96-100 °C

SMILES string

COc1ccc(cc1)C(=O)CCl

InChI

1S/C9H9ClO2/c1-12-8-4-2-7(3-5-8)9(11)6-10/h2-5H,6H2,1H3

InChI key

MCRINSAETDOKDE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service