Skip to Content
MilliporeSigma
All Photos(1)

Documents

716200

Sigma-Aldrich

4-Aminophenylboronic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H8BNO2 · HCl
CAS Number:
Molecular Weight:
173.41
MDL number:
UNSPSC Code:
12352103

mp

195-200 °C

SMILES string

Cl.Nc1ccc(cc1)B(O)O

InChI

1S/C6H8BNO2.ClH/c8-6-3-1-5(2-4-6)7(9)10;/h1-4,9-10H,8H2;1H

InChI key

QBYGJJSFMOVYOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride

Application

Reactant for:
  • Preparation of chromen-4-ones as DNA-dependent protein kinase inhibitors

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service