Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

715565

Sigma-Aldrich

Benzo[c][1,2,5]thiadiazol-5-ylboronic acid pinacol ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15BN2O2S
CAS Number:
Molecular Weight:
262.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-86 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc3nsnc3c2

InChI

1S/C12H15BN2O2S/c1-11(2)12(3,4)17-13(16-11)8-5-6-9-10(7-8)15-18-14-9/h5-7H,1-4H3

InChI key

KISHNZJGTMYYKH-UHFFFAOYSA-N

Application

Benzo[c][1,2,5]thiadiazol-5-ylboronic acid pinacol ester can be used as a reactant:
  • To synthesize 5-methylbenzo[c][1,2,5]thiadiazole by methylation reaction with methyl iodide using palladium catalyst.
  • In the Miyaura borylation and Suzuki coupling reactions.
  • To prepare benzothiadiazole derivatives as potent PFKFB3 kinase inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and Structure-Activity Relationships of N-Aryl 6-Aminoquinoxalines as Potent PFKFB3 Kinase Inhibitors
Boutard N, et al.
ChemMedChem, 14(1), 169-181 (2019)
A Monophosphine Ligand Derived from Anthracene Photodimer: Synthetic Applications for Palladium-Catalyzed Coupling Reactions
Wang X, et al.
Organic Letters, 21(20), 8158-8163 (2019)
Palladium-Catalyzed Methylation of Aryl, Heteroaryl, and Vinyl Boronate Esters
Haydl AM and Hartwig JF
Organic Letters, 21(5), 1337-1341 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service