Skip to Content
MilliporeSigma
All Photos(1)

Documents

715328

Sigma-Aldrich

2-Fluoro-5-formylphenylboronic acid pinacol ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BFO3
CAS Number:
Molecular Weight:
250.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

35-40 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2cc(C=O)ccc2F

InChI

1S/C13H16BFO3/c1-12(2)13(3,4)18-14(17-12)10-7-9(8-16)5-6-11(10)15/h5-8H,1-4H3

InChI key

WHYMLQVABFAGEK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service