Skip to Content
MilliporeSigma
All Photos(2)

Documents

715301

Sigma-Aldrich

Manganese bis(trifluoromethanesulfonate)

Synonym(s):

Manganese triflate, Manganese trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2F6MnO6S2
CAS Number:
Molecular Weight:
353.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: manganese
reagent type: catalyst

impurities

5% metallic Mn

mp

>400 °C

SMILES string

FC(F)(F)S(=O)(=O)O[Mn]OS(=O)(=O)C(F)(F)F

InChI

1S/2CHF3O3S.Mn/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

HEYNLDRKZOOEDN-UHFFFAOYSA-L

General description

Manganese bis(trifluoromethanesulfonate) is also known as manganese triflate. It is used as a catalyst in organic synthesis for the oxidation of various hydrocarbons, epoxidation, and C-H oxidation reactions.

Application

Catalyst for:
  • Biginelli reactions
  • Methanolysis

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service