Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

714739

Sigma-Aldrich

5-Cyano-1,3-benzenedicarboxylic acid

greener alternative

≥95%

Synonym(s):

5-Cyanoisophthalic acid, H2CyIPT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5NO4
CAS Number:
Molecular Weight:
191.14
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

248-252 °C

greener alternative category

SMILES string

OC(=O)c1cc(cc(c1)C(O)=O)C#N

InChI

1S/C9H5NO4/c10-4-5-1-6(8(11)12)3-7(2-5)9(13)14/h1-3H,(H,11,12)(H,13,14)

InChI key

YKADUTAIRWMMFI-UHFFFAOYSA-N

General description

5-Cyano-1,3-benzenedicarboxylic acid (CYDC) is an organic linker that can be used as a ligand in the formation of metal-organic frameworks (MOFs). It is reported to show luminescence in the solid-state at λem= 360 and 372 nm and a small band at 481 nm upon excitation at λexc= 320 nm.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

CYDC is a carboxylate ligand that can be used in the development of new materials for applications in light-emitting diodes (LEDs) and chemical sensors. It can also be used in the formation of multivariate MOFs for potential usage in gas storage, catalysis, and biomedicine.
Metal organic frameworks (MOFs) may be formed by the hydrothermal reaction of 5-Cyano-1,3-benzenedicarboxylic acid ligand with metallic salts of cadmium, zinc and cobalt in the presence of sodium azide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Slow relaxation of magnetization in 3D-MOFs based on dysprosium dinuclear entities bridged by dicarboxylic linkers
Oyarzabal I, et al.
CrystEngComm, 18(17), 3055-3063 (2016)
Luminescence and magnetic properties of three metal?organic frameworks based on the 5-(1H-tetrazol-5-yl) isophthalic acid ligand
Calahorro A, et al.
CrystEngComm, 15(38), 7636-7639 (2013)
Designing Multifunctional 5-Cyanoisophthalate-Based Coordination Polymers as Single-Molecule Magnets, Adsorbents, and Luminescent Materials
Seco JM, et al.
Inorganic Chemistry, 55(21), 11230-11248 (2016)
Multi-Spectroscopic Interrogation of the Spatial Linker Distribution in Defect-Engineered Metal-Organic Framework Crystals: The [Cu3 (btc) 2- x (cydc) x] Showcase
Rivera-Torrente M, et al.
Chemistry?A European Journal null

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service