Skip to Content
MilliporeSigma
All Photos(1)

Documents

714461

Sigma-Aldrich

1-Ethoxycarbonyl cyclobutane-1-carboxylic acid

95%

Synonym(s):

Cyclobutane-1,1-dicarboxylic acid monoethyl ester, Monoethyl 1,1-cyclobutanedicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O4
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.451

density

1.117 g/mL at 25 °C

SMILES string

CCOC(=O)C1(CCC1)C(O)=O

InChI

1S/C8H12O4/c1-2-12-7(11)8(6(9)10)4-3-5-8/h2-5H2,1H3,(H,9,10)

InChI key

XYHZRSFKDSWLHW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service