Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712892

Sigma-Aldrich

(S)-(−)-2-(Dibenzylamino)propionaldehyde

97%

Synonym(s):

(S)-2-(Dibenzylamino)propanal, N,N-Dibenzyl-L-alaninal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO
CAS Number:
Molecular Weight:
253.34
Beilstein:
4293933
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Assay

96.5-103.5% (NT)
97%

form

crystals

optical activity

[α]/D -36.0±2.0°, c = 1 in ethanol

functional group

aldehyde
amine
phenyl

storage temp.

2-8°C

SMILES string

C[C@@H](C=O)N(Cc1ccccc1)Cc2ccccc2

InChI

1S/C17H19NO/c1-15(14-19)18(12-16-8-4-2-5-9-16)13-17-10-6-3-7-11-17/h2-11,14-15H,12-13H2,1H3/t15-/m0/s1

InChI key

GFYXFRCVQSKSDO-HNNXBMFYSA-N

Application

(S)-(-)-2-(Dibenzylamino)propionaldehyde can be employed as a building block for the preparation of:
  • Linezolid dipeptide derivatives and 3-oxazolidin-2-one analogs.
  • (2R,3S)-3-Dibenzylamine-1-nitrobutan-2-ol by reacting with bromonitromethane via nitro-aldol reaction using SmI2 as a catalyst.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and evaluation of the antibacterial activity of new Linezolid dipeptide-type analogues
Garcia-Olaiz GD, et al.
Bioorganic Chemistry, 95, 103483-103483 (2020)
Antibacterial activity of new oxazolidin-2-one analogues in methicillin-resistant Staphylococcus aureus strains
Cordova-Guerrero, et al.
International Journal of Molecular Sciences, 15(4), 5277-5291 (2014)
Efficient nitro-aldol reaction using SmI2: A new route to nitro alcohols under very mild conditions
Concellon J, et al.
The Journal of Organic Chemistry, 71(20), 7919-7922 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service