Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712884

Sigma-Aldrich

(S)-(+)-Methyl indoline-2-carboxylate

≥97.0% (HPLC)

Synonym(s):

(S)-2,3-Dihydro-1H-indole-2-carboxylic acid methyl ester, (S)-Indoline-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Beilstein:
5334940
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

crystals

optical activity

[α]/D 31.0±1.5°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H]1Cc2ccccc2N1

InChI

1S/C10H11NO2/c1-13-10(12)9-6-7-4-2-3-5-8(7)11-9/h2-5,9,11H,6H2,1H3/t9-/m0/s1

InChI key

URORFKDEPJFPOV-VIFPVBQESA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service