Skip to Content
MilliporeSigma
All Photos(1)

Documents

712736

Sigma-Aldrich

6-Azido-6-deoxy-1,2:3,4-di-O-isopropylidene-α-D-galactopyranose

≥96% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19N3O5
CAS Number:
Molecular Weight:
285.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96% (HPLC)

form

liquid

optical activity

[α]/D -92.0±3.0°, c = 1 in ethyl acetate

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

CC1(C)O[C@H]2O[C@H](CN=[N+]=[N-])[C@@H]3OC(C)(C)O[C@@H]3[C@H]2O1

InChI

1S/C12H19N3O5/c1-11(2)17-7-6(5-14-15-13)16-10-9(8(7)18-11)19-12(3,4)20-10/h6-10H,5H2,1-4H3/t6-,7+,8+,9-,10-/m1/s1

InChI key

KIBLVBPHQCVUFG-SOYHJAILSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service