Skip to Content
MilliporeSigma

71270

Sodium amide-sodium tert-butylate complex base solution

technical, ~0.8 M in THF (suspension)

Synonym(s):

Complex base from sodium amide/Sodium tert-butoxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
2NaNH2 · (CH3)3CONa
CAS Number:
Molecular Weight:
174.13
UNSPSC Code:
12352301
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical

concentration

~0.8 M in THF (suspension)

density

0.92 g/mL at 20 °C

SMILES string

[NH2-].[NH2-].[Na+].[Na+].[Na+].CC(C)(C)[O-]

Other Notes

Complex base employed to generate carbanions




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library