Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712590

Sigma-Aldrich

O-(2-Azidoethyl)-O′-methyl-triethylene glycol

≥90% (NMR)

Synonym(s):

1-{2-[2-(2-Azidoethoxy)ethoxy]ethoxy}-2-methoxyethane, 13-Azido-2,5,8,11-tetraoxatridecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19N3O4
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90% (NMR)

form

liquid

mol wt

average Mn 200

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

Ω-end

azide

α-end

methoxy

storage temp.

2-8°C

SMILES string

COCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C9H19N3O4/c1-13-4-5-15-8-9-16-7-6-14-3-2-11-12-10/h2-9H2,1H3

InChI key

FFOZZVDSANUDAE-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service