Skip to Content
MilliporeSigma
All Photos(1)

Documents

712051

Sigma-Aldrich

L-α-Cyclopropylglycine

95%

Synonym(s):

(S)-Amino-cyclopropylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
5728866
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

94.0-106.0% (NT)
95%

form

lumps

optical activity

[α]/D +101±4°, c = 1% in 1 M HCl

impurities

≤5.0% water

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@@H](C1CC1)C(O)=O

InChI

1S/C5H9NO2/c6-4(5(7)8)3-1-2-3/h3-4H,1-2,6H2,(H,7,8)/t4-/m0/s1

InChI key

BUSBCPMSNBMUMT-BYPYZUCNSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service