Skip to Content
MilliporeSigma
All Photos(2)

Documents

711322

Sigma-Aldrich

Chlorocarbonylgold(I)

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CAuClO
CAS Number:
Molecular Weight:
260.43
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

247-253 °C (dec.)

storage temp.

−20°C

SMILES string

Cl[Au].[C-]#[O+]

InChI

1S/CO.Au.ClH/c1-2;;/h;;1H/q;+1;/p-1

InChI key

VBUSSYWEWHQPTL-UHFFFAOYSA-M

Application

Reactant for silylene insertion into gold-chlorine bonds

Used for stabilization and encapsulation inside functionalized zeolite imidazolate frameworks

Transition-metal-catalyst for oxidation of carbon monoxide by dichlorine to produce phosgene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service