Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710539

Sigma-Aldrich

Bis(isopropylcyclopentadienyl)nickel

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22Ni
CAS Number:
Molecular Weight:
273.04
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

liquid

reaction suitability

reagent type: catalyst
core: nickel

density

1.129 g/mL at 25 °C
1.15 g/mL at 25 °C

storage temp.

−20°C

SMILES string

[Ni].CC(C)[C]1[CH][CH][CH][CH]1.CC(C)[C]2[CH][CH][CH][CH]2

InChI

1S/2C8H11.Ni/c2*1-7(2)8-5-3-4-6-8;/h2*3-7H,1-2H3;

InChI key

OIYDMFLWYJTIRX-UHFFFAOYSA-N

Application

Phosphine ligand for cross coupling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service