Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710504

Sigma-Aldrich

Bis(4-methoxyphenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15O2P
CAS Number:
Molecular Weight:
246.24
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reagent type: ligand

mp

36-40 °C

functional group

phosphine

SMILES string

COc1ccc(Pc2ccc(OC)cc2)cc1

InChI

1S/C14H15O2P/c1-15-11-3-7-13(8-4-11)17-14-9-5-12(16-2)6-10-14/h3-10,17H,1-2H3

InChI key

FWKICRREDMVWRF-UHFFFAOYSA-N

Application

Reactant for:
  • Enantioselective Ir-catalyzed hydrogenation
  • Palladium-catalyzed asymmetric additions
  • Alkali-metal-catalyzed addition reactions
  • Preparation of different DIOP-type ligands for uses in Rh-catalyzed asymmetric hydrogenation

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service