Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710415

Sigma-Aldrich

Bis(isopropylcyclopentadienyl)titanium dichloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22Cl2Ti
CAS Number:
Molecular Weight:
333.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Quality Level

Assay

95%

form

solid

reaction suitability

core: titanium
reagent type: catalyst

mp

171-173 °C

SMILES string

Cl[Ti]Cl.CC(C)[C]1[CH][CH][CH][CH]1.CC(C)[C]2[CH][CH][CH][CH]2

InChI

1S/2C8H11.2ClH.Ti/c2*1-7(2)8-5-3-4-6-8;;;/h2*3-7H,1-2H3;2*1H;/q;;;;+2/p-2

InChI key

HMTIYOSDNUJYPV-UHFFFAOYSA-L

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service