Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710032

Sigma-Aldrich

Cyclohexylboronic acid MIDA ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18BNO4
CAS Number:
Molecular Weight:
239.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

210-215 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)C2CCCCC2

InChI

1S/C11H18BNO4/c1-13-7-10(14)16-12(17-11(15)8-13)9-5-3-2-4-6-9/h9H,2-8H2,1H3

InChI key

SLUNFPHQHGXTCG-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service