Skip to Content
MilliporeSigma
All Photos(1)

Documents

709069

Sigma-Aldrich

Hexammine cobalt(III) chloride-d18

95 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Co(ND3)6]Cl3
CAS Number:
Molecular Weight:
285.59
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

95 atom % D

Assay

95% (CP)

form

solid

mol wt

284.68 by atom % calculation

mp

217 °C

mass shift

M+18

SMILES string

[2H]N([2H])[2H].[2H]N([2H])[2H].[2H]N([2H])[2H].[2H]N([2H])[2H].[2H]N([2H])[2H].[2H]N([2H])[2H].Cl[Co](Cl)Cl

InChI

1S/3ClH.Co.6H3N/h3*1H;;6*1H3/q;;;+3;;;;;;/p-3/i/hD18

InChI key

JXBGZYGSWFSYFI-DLGCUCRUSA-K

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service