Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708593

Sigma-Aldrich

1,3-Dimethyl-1H-benzimidazolium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2
CAS Number:
Molecular Weight:
274.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: catalyst

mp

137-142 °C

SMILES string

[I-].Cn1c[n+](C)c2ccccc12

InChI

1S/C9H11N2.HI/c1-10-7-11(2)9-6-4-3-5-8(9)10;/h3-7H,1-2H3;1H/q+1;/p-1

InChI key

RQGURHMTNSNBQX-UHFFFAOYSA-M

Application

Catalyst for:
  • Domino ring-opening redox amidation Knoevenagel condensation
  • Intramolecular stereoselective protonation
  • Grignard allylic substitution
  • Acylation of alcohols
  • Umpolung reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service