Skip to Content
MilliporeSigma
All Photos(1)

Documents

708550

Sigma-Aldrich

(1R,2R)-N,N′-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)

≥97%

Synonym(s):

(R,R)-AMAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H38CoN2O4
CAS Number:
Molecular Weight:
573.59
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

solid

optical activity

[α]/D 291°, c = 0.1 in chloroform

mp

246-250 °C

SMILES string

CC(=O)C1=C(C)O[Co]OC(C)=C(\C=N\[C@@H]([C@H](\N=C\1)c2c(C)cc(C)cc2C)c3c(C)cc(C)cc3C)C(C)=O

InChI

1S/C32H40N2O4.Co/c1-17-11-19(3)29(20(4)12-17)31(33-15-27(23(7)35)24(8)36)32(34-16-28(25(9)37)26(10)38)30-21(5)13-18(2)14-22(30)6;/h11-16,31-32,35,37H,1-10H3;/q;+2/p-2/b27-23+,28-25+,33-15+,34-16+;/t31-,32-;/m1./s1

InChI key

PHCQQLMRNZRDJA-ZQBGJXBSSA-L

Application

  • Schiff base-cobalt complex catalyst for asymmetric borohydride reduction of carbonyl compounds

Catalyst for:
  • Enantioselective borohydride reduction
  • Stereoselective preparation of aryl alcohols via asymmetric reduction of aryl ketones with borohydride
  • Stereoselective borohydride reduction of diketones to diols
  • Stereoselective preparation of optically active deuterated primary alcohols via enantioselective borodeuteride reduction of aldehydes
  • Stereoselective preparation of β-hydroxy esters via dynamic kinetic resolution of alkylketo esters with enantioselective borohydride reduction
  • Chemoselective, diastereoselective, and enantioselective borohydride reduction of 1,3-diketones
  • Asymmetric synthesis of diarylhydroxypropanones by stereoselective reduction of alkyldiaryldiketones
Catalyst used for the enantioselective reduction of ketones, hetero Diels-Alder reaction, carbonylene reaction, 1,3-dipolar cycloaddition

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yamada, T.
Synthesis, 10, 1628-1628 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service