Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708038

Sigma-Aldrich

2,3-Butanedione-1,4-13C2

99 atom % 13C, 97% (CP)

Synonym(s):

Biacetyl-1,4-13C2, Diacetyl-1,4-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3COCO13CH3
CAS Number:
Molecular Weight:
88.07
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

bp

88 °C

density

1.003 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

[13CH3]C(=O)C([13CH3])=O

InChI

1S/C4H6O2/c1-3(5)4(2)6/h1-2H3/i1+1,2+1

InChI key

QSJXEFYPDANLFS-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service