Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

707929

Sigma-Aldrich

4-Isoquinolineboronic acid

Synonym(s):

Isoquinolin-4-ylboronic acid, Isoquinoline-4-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BNO2
CAS Number:
Molecular Weight:
172.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

Quality Level

mp

230-234 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cncc2ccccc12

InChI

1S/C9H8BNO2/c12-10(13)9-6-11-5-7-3-1-2-4-8(7)9/h1-6,12-13H

InChI key

GDTOUTKTCGPAGY-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Application

Reactant for:
  • Preparation of selective steroid-11β-hydroxylase (CYP11B1) inhibitors for treatment of cortisol dependent diseases
  • Preparation of tetrabutylammonium trifluoroborates
  • Preparation of heteroaryl substituted tetrahydropyrroloijquinolinone derivatives as aldosterone synthase inhibitors
  • Synthesis of aminoarylpyridazines as selective CB2 agonists for treatment of inflammatory pain
  • Preparation of acyl substituted indoles via palladium-catalyzed domino coupling/carbonylation/Suzuki coupling of dibromoethenylanilines
  • Synthesis of antagonists of bacterial quorum sensing
  • Preparation of potassium heteroaryl trifluoroborates from boronic acids and their use as coupling partners with various aryl and heteroaryl halides in Suzuki-Miyaura cross-coupling reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service