Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

707481

Sigma-Aldrich

Polyhydroxy small gap fullerenes, hydrated

Synonym(s):

Fullerenols, Polyhydroxy fullerenes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C120O30(OH)30 · 30H2O · 25Na+
UNSPSC Code:
12352103

form

crystals

Quality Level

composition

Polyhydroxy SGFs(TGA), ~85%

mp

60 °C (dec.)

solubility

water, pH >9: soluble

SMILES string

O[C]12=C3c4c5c6c7c8c9c%10c%11c%12c%13c%14c%15c%16c(C3=[C]%17(O)c%18c1c%19c%20c%21c%22c%23c%24c%25c%26c%27c%28c%29c(C%17=[C]%16(O)[C]%14(O)=C%29[C]%27(O)=C%13[C]%11(O)=C%26[C]%24(O)=C%10[C]8(O)=C%23[C]%21(O)=C7[C]5(O)=C2%20)c%18c%30c%28c%25c%22c%19%30)c4c%31c6c9c%12c%15%31

General description

The starting material, small gap fullerenes, ranges from C60 up to C400 with the peak distribution between C100 and C120 in TOF-MS.

Application

Interaction of hydrated polyhydroxy small gap fullerenes with Caco-2 was studied. The nanomaterial can find potential use as drug delivery agent.

Preparation Note

The starting material, small gap fullerenes, ranges from C60 up to C400 with the peak distribution between C100 and C120 in TOF-MS.

Other Notes

Structure image is a representative fullerene, this product is heterogeneous.

Legal Information

Product of TDA Research, Inc. and US Patents 6,517,799 and 6,303,016.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical redox recovery of giant, small-gap and other fullerenes
Raebiger, J.W.; Alford, J.M.; Bolskar, R.D.; Diener, M.D.
Carbon, 49, 37-46 (2011)
Functionalized carbon nanomaterials: exploring the interactions with Caco-2 cells for potential oral drug delivery
Coyuco JC, et al.
International journal of nanomedicine, 6, 2253-2253 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service