Skip to Content
MilliporeSigma
All Photos(1)

Documents

706639

Sigma-Aldrich

N-Fmoc-3-piperidinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO3
CAS Number:
Molecular Weight:
321.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

powder

mp

90-96 °C

functional group

Fmoc

SMILES string

O=C1CCCN(C1)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C20H19NO3/c22-14-6-5-11-21(12-14)20(23)24-13-19-17-9-3-1-7-15(17)16-8-2-4-10-18(16)19/h1-4,7-10,19H,5-6,11-13H2

InChI key

ZHMRQZFSADRNCI-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service