Skip to Content
MilliporeSigma
All Photos(1)

Documents

706612

Sigma-Aldrich

2-Bromo-3-(dimethylamino)acrolein

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8BrNO
CAS Number:
Molecular Weight:
178.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

67-72 °C

storage temp.

−20°C

SMILES string

CN(C)\C=C(/Br)C=O

InChI

1S/C5H8BrNO/c1-7(2)3-5(6)4-8/h3-4H,1-2H3/b5-3-

InChI key

ACBSCWFIKSDJLP-HYXAFXHYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service