Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706485

Sigma-Aldrich

4,6-Dichloro-5-methoxypyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Cl2N2O
CAS Number:
Molecular Weight:
179.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

54-58 °C

functional group

chloro

SMILES string

COc1c(Cl)ncnc1Cl

InChI

1S/C5H4Cl2N2O/c1-10-3-4(6)8-2-9-5(3)7/h2H,1H3

InChI key

IJQIGKLDBGKSNT-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonah H Mwalwisi et al.
Journal of pharmaceutical and biomedical analysis, 129, 558-570 (2016-08-10)
A simple, cost effective, accurate, and precise RP-HPLC method was developed for the simultaneous determination of sulfalene and sulfadoxine in fixed dose dual combinations with pyrimethamine together with their related substances. Proprietary products containing these combinations are often being prescribed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service