Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706477

Sigma-Aldrich

N-Fmoc-piperidine-2-carboxylic acid

97%

Synonym(s):

1,2-Piperidinedicarboxylic acid, 1-(9H-fluoren-9-ylmethyl) ester, Fmoc-DL-pipecolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

113-120 °C

application(s)

peptide synthesis

functional group

Fmoc
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(=O)C1CCCCN1C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C21H21NO4/c23-20(24)19-11-5-6-12-22(19)21(25)26-13-18-16-9-3-1-7-14(16)15-8-2-4-10-17(15)18/h1-4,7-10,18-19H,5-6,11-13H2,(H,23,24)

InChI key

CKLAZLINARHOTG-UHFFFAOYSA-N

Application

Reactant for racemic solid-phase preparation of human protease activated receptor 2 agonists and antagonists

Analyte for urea-linked cinchona-calexarene hybrid type receptors for chromatographic enantiomer separation of carbamate-protected amino acids

Combinatorial reactant for assembly of alkylglycine library arrays by SPOT technique on cellulose membranes for identification of micromolar ligands for monoclonal antibodies

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service