Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706116

Sigma-Aldrich

Potassium benzofurazan-5-trifluoroborate

96%

Synonym(s):

Potassium benzo[c][1,2,5]oxadiazole-5-trifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BF3KN2O
CAS Number:
Molecular Weight:
226.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

>300 °C

SMILES string

[K+].F[B-](F)(F)c1ccc2nonc2c1

InChI

1S/C6H3BF3N2O.K/c8-7(9,10)4-1-2-5-6(3-4)12-13-11-5;/h1-3H;/q-1;+1

InChI key

VVXGHNBCIPLVLI-UHFFFAOYSA-N

Application

Organotrifluoroborate invovled in copper-mediated cross-coupling and Suzuki cross-coupling

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 5-aryl- and 5-heteroaryl-7-carboxyl-8-hydroxyquinaldines through Suzuki cross-coupling reaction with potassium organotrifluoroborates
F. Sliman and D. Desmaele,
Synthesis, 4, 619-630 (2010)
Sebastian A Testero et al.
Bioorganic & medicinal chemistry letters, 21(9), 2675-2678 (2011-01-25)
The copper-mediated and non-basic oxidative cross-coupling of organotrifluoroborates with phenols was applied to elaboration of the structures of thiirane-based inhibitors of matrix metalloproteinases. By revision of the synthetic sequence to allow this cross-coupling as the final step, and taking advantage

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service