Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

705179

Sigma-Aldrich

N,N-Diisopropyl-4H-benzo[d][1,3,2]dioxaborinin-2-amine

97%

Synonym(s):

(4H-Benzo[d][1,3,2]dioxaborinin-2-yl)-diisopropyl-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20BNO2
CAS Number:
Molecular Weight:
233.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.505

density

1.020 g/mL at 25 °C

SMILES string

CC(C)N(B1OCc2ccccc2O1)C(C)C

InChI

1S/C13H20BNO2/c1-10(2)15(11(3)4)14-16-9-12-7-5-6-8-13(12)17-14/h5-8,10-11H,9H2,1-4H3

InChI key

LBKWVNXGUHDPET-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.0 °F

Flash Point(C)

62.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service