Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

705160

Sigma-Aldrich

3,5,6,8-Tetrabromo-1,10-phenanthroline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Br4N2
CAS Number:
Molecular Weight:
495.79
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

SMILES string

Brc1cnc2c(c1)c(Br)c(Br)c3cc(Br)cnc23

InChI

1S/C12H4Br4N2/c13-5-1-7-9(15)10(16)8-2-6(14)4-18-12(8)11(7)17-3-5/h1-4H

InChI key

BAPJVWLAFSQLNX-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service