Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704482

Sigma-Aldrich

Methyl 2-aminopyridine-4-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

144-148 °C

functional group

ester

SMILES string

COC(=O)c1ccnc(N)c1

InChI

1S/C7H8N2O2/c1-11-7(10)5-2-3-9-6(8)4-5/h2-4H,1H3,(H2,8,9)

InChI key

SVWWNEYBEFASMP-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesús López-Molina et al.
Dalton transactions (Cambridge, England : 2003), 49(14), 4315-4322 (2020-03-13)
This study tries to provide new solutions to increase the efficiency of conversion of photons in solar cells, using photoluminescent Cu(i) coordination polymers (CPs) as possible alternative materials of lower cost, than those used today, based on lanthanides. The selected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service