Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704377

Sigma-Aldrich

Tris(hydroxymethyl-13C)nitromethane

99 atom % 13C, 98% (CP)

Synonym(s):

2-(Hydroxymethyl-13C)-2-nitro-1,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO13CH2)3CNO2
CAS Number:
Molecular Weight:
154.10
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

technique(s)

NMR: suitable

mp

160 °C (dec.)

mass shift

M+3

SMILES string

O[13CH2]C([13CH2]O)([13CH2]O)[N+]([O-])=O

InChI

1S/C4H9NO5/c6-1-4(2-7,3-8)5(9)10/h6-8H,1-3H2/i1+1,2+1,3+1

InChI key

OLQJQHSAWMFDJE-VMIGTVKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service