Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

703850

Sigma-Aldrich

Ethyl cyano-13C, 15N-acetate-1,2-13C2

99 atom % 13C, 98 atom % 15N, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15N≡13C13CH213COOCH2CH3
CAS Number:
Molecular Weight:
117.09
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

98% (CP)

form

liquid

bp

207-210 °C

density

1.100 g/mL at 25 °C

mass shift

M+4

storage temp.

2-8°C

SMILES string

CCO[13C](=O)[13CH2][13C]#[15N]

InChI

1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3/i3+1,4+1,5+1,6+1

InChI key

ZIUSEGSNTOUIPT-PQVJJBODSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F

Flash Point(C)

109 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service