Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

702951

Sigma-Aldrich

Cy-cBRIDP

97%

Synonym(s):

1-(Dicyclohexylphosphino)-2,2-Diphenyl-1-methylcyclopropane, Dicyclohexyl(2,2-diphenyl-1-methyl-1-cyclopropyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37P
CAS Number:
Molecular Weight:
404.57
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

115-122 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

CC1(CC1(c2ccccc2)c3ccccc3)P(C4CCCCC4)C5CCCCC5

InChI

1S/C28H37P/c1-27(29(25-18-10-4-11-19-25)26-20-12-5-13-21-26)22-28(27,23-14-6-2-7-15-23)24-16-8-3-9-17-24/h2-3,6-9,14-17,25-26H,4-5,10-13,18-22H2,1H3

InChI key

JDNAVZKVTWHWKB-UHFFFAOYSA-N

Legal Information

Sold in collaboration with Takasago for research purposes only. US7129367

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service