Skip to Content
MilliporeSigma
All Photos(1)

Documents

702234

Sigma-Aldrich

Lithium-6Li nitrate

95 atom % 6Li, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
6LiNO3
CAS Number:
Molecular Weight:
68.02
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

95 atom % 6Li

Assay

98% (CP)

form

powder

mp

264 °C (lit.)

mass shift

M-1

SMILES string

[6Li+].[O-][N+]([O-])=O

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1/i1-1;

InChI key

IIPYXGDZVMZOAP-ULWFUOSBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service