Skip to Content
MilliporeSigma
All Photos(1)

Documents

701777

Sigma-Aldrich

Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate

Synonym(s):

5-2,4-Cyclopentadien-1-yl)[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H19Ru · PF6
CAS Number:
Molecular Weight:
445.35
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

83-84 °C

SMILES string

[Ru+].[CH]1[CH][CH][CH][CH]1.F[P-](F)(F)(F)(F)F.CC(C)c2ccc(C)cc2

InChI

1S/C10H14.C5H5.F6P.Ru/c1-8(2)10-6-4-9(3)5-7-10;1-2-4-5-3-1;1-7(2,3,4,5)6;/h4-8H,1-3H3;1-5H;;/q;;-1;+1

InChI key

KHUJNZPWTSOTDJ-UHFFFAOYSA-N

Application

Reactant for:
  • Synthesis of ruthenocenium complexes
  • Intercalation reactions

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service