Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70170

Sigma-Aldrich

Naphazoline nitrate

≥99.0% (NT)

Synonym(s):

2-(1-Naphthylmethyl)imidazoline nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2 · HNO3
CAS Number:
Molecular Weight:
273.29
Beilstein:
3779329
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0% (NT)

form

crystals

ign. residue

≤0.1%

loss

≤0.5% loss on drying

mp

167-170 °C

SMILES string

O[N+]([O-])=O.C1CN=C(Cc2cccc3ccccc23)N1

InChI

1S/C14H14N2.HNO3/c1-2-7-13-11(4-1)5-3-6-12(13)10-14-15-8-9-16-14;2-1(3)4/h1-7H,8-10H2,(H,15,16);(H,2,3,4)

InChI key

ZAHXYMFVNNUHCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Therapeutic use to:
  • Combat ammonia poisoning
  • Treat seasonal allergy rhinitis

Involved in studies on the effects of imidazole drugs on chronotropic and inotropic effects

Pharmacological activity for topical mydriatics and vasoconstrictors

Radioprotector for mice treated with ionizing radiation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service