Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

701327

Sigma-Aldrich

3-Formyl-1H-indole-5-carbonitrile

97%

Synonym(s):

3-Formyl-5-cyano-1H-indole, 5-Cyanoindole-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2O
CAS Number:
Molecular Weight:
170.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

248-253 °C

functional group

aldehyde
nitrile

SMILES string

O=Cc1c[nH]c2ccc(cc12)C#N

InChI

1S/C10H6N2O/c11-4-7-1-2-10-9(3-7)8(6-13)5-12-10/h1-3,5-6,12H

InChI key

NVBCFOQYDFKXJJ-UHFFFAOYSA-N

Application

  • Reactant for synthesis of tryptophan dioxygenase inhibitors as potential anticancer immunomodulators
  • Reactant for enantioselective preparation of antifungal agents
  • Reactant for synthesis of indole- and 7-azaindole-1,3-dicarboxamide hydroxyethylamine inhibitors of BACE-1
  • Reactant for preparation of indolyl alkenes from microwave-enhanced Knoevenagel condensation as antibacterial agents
  • Reactant for preparation of indolecarboxamide derivatives as antitumor agents
  • Reactant for synthesis of a selective serotonin reuptake inhibitor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service