Skip to Content
MilliporeSigma
All Photos(1)

Documents

700711

Sigma-Aldrich

N-(Dicyclohexylphosphino)-2-(2′-tolyl)indole

95%

Synonym(s):

N-(dicyclohexylphosphino)-2-(2′-methylphenyl)-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34NP
CAS Number:
Molecular Weight:
403.54
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

139-145 °C

functional group

phosphine

SMILES string

Cc1ccccc1-c2cc3ccccc3n2P(C4CCCCC4)C5CCCCC5

InChI

1S/C27H34NP/c1-21-12-8-10-18-25(21)27-20-22-13-9-11-19-26(22)28(27)29(23-14-4-2-5-15-23)24-16-6-3-7-17-24/h8-13,18-20,23-24H,2-7,14-17H2,1H3

InChI key

KESYNFLRGDUANT-UHFFFAOYSA-N

Application

Catalyst for Suzuki-Miyaura coupling reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service