Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700231

Sigma-Aldrich

Ethynylboronic acid MIDA ester

Synonym(s):

Acetyleneboronic acid MIDA ester, Acetynylboronic acid MIDA ester, Ethyneboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BNO4
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

208-209 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)C#C

InChI

1S/C7H8BNO4/c1-3-8-12-6(10)4-9(2)5-7(11)13-8/h1H,4-5H2,2H3

InChI key

NHVZIRZCTRJEFP-UHFFFAOYSA-N

Application

Ethynylboronic acid MIDA ester is a bifunctional acetylene equivalent, air-stable, chromatography-compatible boronate commonly used in Suzuki-Miyaura cross-coupling reaction. It can also be used in the synthesis of heterocyclic MIDA boronates of isoquinolone, isoquinoline, pyrrole, indole, isoxazoles and triazoles for subsequent cross-coupling reactions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethynyl MIDA boronate: a readily accessible and highly versatile building block for small molecule synthesis.
Struble J R, et al.
Tetrahedron, 66(26), 4710-4718 (2010)
Regioselective synthesis and slow-release Suzuki?Miyaura cross-coupling of MIDA boronate-functionalized isoxazoles and triazoles.
Grob J E, et al.
The Journal of Organic Chemistry, 76(24), 10241-10248 (2011)
Mild Rh (III)-catalyzed C?H activation and annulation with alkyne mida boronates: Short, efficient synthesis of heterocyclic boronic acid derivatives.
Wang H, et al.
Journal of the American Chemical Society, 134(48), 19592-19595 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service