Skip to Content
MilliporeSigma
All Photos(2)

Documents

699888

Sigma-Aldrich

1,8-Octanediphosphonic acid

97%

Synonym(s):

1,8-Diphosphonooctane, 1,8-octanediylbis-phosphonic acid, C8BPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20O6P2
CAS Number:
Molecular Weight:
274.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

180-185 °C

SMILES string

OP(O)(=O)CCCCCCCCP(O)(O)=O

InChI

1S/C8H20O6P2/c9-15(10,11)7-5-3-1-2-4-6-8-16(12,13)14/h1-8H2,(H2,9,10,11)(H2,12,13,14)

InChI key

VRAVNKVHQXXAQW-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service